Restek™ Straight Inlet Liners for Splitless Injection
Used for samples containing a narrow molecular weight distribution and for those not prone to thermal decomposition
Manufacturer: Restek™ 20712
 View more versions of this product
Catalog No. 06-710-932
  Check Availability
Add to Cart
    Specifications
    Product Type IP splitless straight inlet liner
    Diameter (Metric) Inner 0.2cm
    Diameter (Metric) Outer 0.65cm
    Length (Metric) 7.85cm
    For Use With (Equipment) Agilent GCs equipped with split/splitless inlets
    Liner Material Borosilicate Glass
    Quantity Each
  • Description & Specifications
Related Products
Product Content Feedback