Learn More
Restek™ Sulfinert Gas Sampling Valves
Sulfinert-treated valves are ideal for samples containing low concentrations of sulfur or other active compounds.
Supplier: Restek™ 20584
Description
- “W Type” Valve
- 0.0625 in. fittings
- 0.40mm Port diameter
Specifications
Sulfinert Coated 4-Port Manual Sample Valve | |
1/16 in. Fittings |