Restek™ Sulfinert-Treated Swagelok Sample Cylinders

Made of 304L stainless steel to resist corrosion and DOT rated to 1800 and 5000psig, which allows sampling at gas wellheads as well as on-site refineries.

Manufacturer: Restek™ 24133

 View more versions of this product

Catalog No. 06-712-203

  Check Availability
Add to Cart

    Specifications

    Material Stainless Steel, Sulfinert™ treated
    Pressure 1800psi
    Size 500cc
    Thread Size (English) 0.25 NPTF in.
    Thread Size (Metric) 0.63 NPTFcm
    Product Type Sample Cylinder

    • Cylinders have 0.25 in. female NPT threads on both ends
    • Each cylinder is hydrostatically tested to at least 5/3 the working pressure
    • Range of cylinder sizes: 75 cc to 2,250 cc

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit