Learn More
Restek™ Super-Clean Gas Trapping System for LC/MS
Quick-change cartridge system removing hydrocarbons efficiently from nitrogen
Supplier: Restek™ 22061
Description
- Changing filters is quick and easy
- Up to 20L of hydrocarbon-free nitrogen per minute
- Filters connected in parallel to handle high flows of LC/MS
Specifications
Hydrocarbon Filter | |
Super-Clean Gas Trapping System |