Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Restek™ Tedlar Sampling Bags
Lightweight and easy to use sampling bags with polypropylene or stainless-steel valves.
Manufacturer:  Restek™ 22039
 View more versions of this product
Catalog No. 06-711-841
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
EPA, NIOSH, OSHA
50cc/m2 x day oxygen, 9 to 57g/m2 x day, water vapor, 172cc/m2 x day carbon dioxide
399.2°F
8000psi
0.05mm
1L
Valve and septum
Tedlar Bag
204°C
0.002 in.
33.81 oz.
Stainless Steel