Learn More
Restek™ Thermolite™ Septa
Precision molding ensures consistent, accurate fit
Manufacturer:  Restek™ 27122
 View more versions of this product
Catalog No. 06-802-702
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
0.196 in.
100/Pk.
5mm
340°C
5mm
Thermolite™ Septa
644°F