Restek™ Thermolite™ Septa (Shimadzu™ Plug)
For SRI GCs
Manufacturer: Restek™ 27154
 View more versions of this product
Catalog No. 06-802-734
  Check Availability
Add to Cart
    Specifications
    Type Thermolite™ Plug Septa
    Size Pack of 50
    Temperature (English) Inlet 644°F
    Temperature (Metric) Inlet 340°C
  • Description & Specifications
Related Products
Product Content Feedback