Restek™ Ultra Cyano HPLC Prep Columns, 5um Particle Size

Easy scale-up from Restek analytical columns.

Manufacturer: Restek™ 9106579

 View more versions of this product

Catalog No. 06-722-385

  Check Availability
Add to Cart

    Specifications

    Packing Material Ultra Cyano
    Stationary Phase Ultra Cyano
    Type HPLC preparative column
    Support Material Silica
    Ligand Type Cyanopropylsilane
    Column Type Ultra Cyano
    Particle Shape Spherical
    Particle Size 5μm
    Pore Size 100Å
    pH 2.5 to 8
    For Use With For normal-phase, reversed-phase (best with mobile phase pH between 5 and 7), and HILIC modes
    Surface Area 300m2/g
    Film Thickness 5μm
    Functionality/Interaction Type Dipole interaction
    Length (Metric) 250mm
    Technique Reversed or normal phase HPLC
    Diameter (Metric) Inner 30mm
    Carbon Load 8%
    Phase Cyano
    Column Format Prep
    Column Chemistry Nitrile groups
    Max. Temperature 80°C
    USP Type L10
    Endcapped Yes

    Restek Cyano is a traditional monomeric cyanopropylsilane that is recommended for assays where alternate selectivity, or confirmation, to a C18 or C8 column is desired. It can be used in normal-phase, reversed-phase and HILIC modes. Ideal for the analysis of protonated bases.
    • Particle Size: 5µm, spherical
    • Pore Size: 100 angstrom
    • Carbon Load: 8%
    • End-Cap: yes
    • pH Range: 2.5 to 8
    • Temperature Limit: 80°C
    • USP Phase Code: L10
    • Phase Category: cyano
    • Ligand Type: cyanopropylsilane

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit