Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Ultra Allure Organic Acids Guard Cartridges
SDP

Requires Trident LC Column Protection System or Trident HPLC In-Line Guard Cartridge Holder

Supplier:  Restek™ 916550212

Catalog No. 06-611-658


Add to Cart

Description

Description

  • Particle Size: 5μm, spherical
  • Pore size: 100 angstrom
Specifications

Specifications

2.1 mm
10 mm
5 μm
3/Pk.
Trident LC Column Protection System or Trident HPLC In-Line Guard Cartridge Holder
Spherical
100 Å
Allure Organic Acids
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit