Learn More
Restek™ Ultra Guard Cartridges
Protects and extends column lifetime.
$191.00 - $233.00
Specifications
Pore Size 100Å
Particle Shape Spherical
pH 2.5 to 8
View More Specs
Products 39
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Quantity Type Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Quantity Type Price Quantity & Availability  
06-716-516
View Documents
Restek™
917750210
4 mm 10 mm Each Ultra Carbamate
Each for $231.00
Add to cart
06-719-064
View Documents
Restek™
910050210
4 mm 10 mm Each Ultra; Silica
Pack of 3 for $231.00
Add to cart
06-719-068
View Documents
Restek™
910150210
4 mm 10 mm Each Ultra C1
Pack of 3 for $228.00
Add to cart
06-719-076
View Documents
Restek™
910350210
4 mm 10 mm Each Ultra C8
Pack of 3 for $233.00
Add to cart
06-719-073
View Documents
Restek™
910250212
2.1 mm 10 mm 3 Pack Ultra C4
Pack of 3 for $228.00
Add to cart
06-719-077
View Documents
Restek™
910350212
3 mm 10 mm 3 Pack Ultra C8
Pack of 3 for $228.00
Add to cart
06-719-084
View Documents
Restek™
910650210
4 mm 10 mm Each Cyano
Pack of 3 for $228.00
Add to cart
06-719-088
View Documents
Restek™
910750210
4 mm 10 mm Each Amino
Pack of 3 for $231.00
Add to cart
06-719-161
View Documents
Restek™
917450210
4 mm 10 mm Each C18
Pack of 3 for $231.00
Add to cart
06-719-162
View Documents
Restek™
917450212
2.1 mm 10 mm 3 Pack C18
Pack of 3 for $231.00
Add to cart
06-719-165
View Documents
Restek™
917550210
4 mm 10 mm Each IBD
Pack of 3 for $228.00
Add to cart
06-719-166
View Documents
Restek™
917550212
2.1 mm 10 mm 3 Pack Ultra; IBD
Pack of 3 for $231.00
Add to cart
06-719-169
View Documents
Restek™
917850210
4 mm 10 mm Each Aqueous C18
Pack of 3 for $229.00
Add to cart
06-719-170
View Documents
Restek™
917850212
4.6 mm 10 mm 3 Pack Aqueous C18
Pack of 3 for $228.00
Add to cart
06-611-657
View Documents
Restek™
912750212
10 mm 3/Pk.
Pack of 3 for $191.00
Add to cart
06-611-656
View Documents
Restek™
912750210
10 mm 3/Pk.
Pack of 3 for $191.00
Add to cart
06-611-655
View Documents
Restek™
910950212
10 mm 3/Pk.
Pack of 3 for $194.00
Add to cart
06-611-654
View Documents
Restek™
910950210
10 mm 3/Pk.
Pack of 3 for $191.00
Add to cart
06-611-659
View Documents
Restek™
917750212
10 mm 3/Pk.
Pack of 3 for $194.00
Add to cart
06-611-653
View Documents
Restek™
910650212
10 mm 3/Pk.
Pack of 3 for $194.00
Add to cart
06-611-651
View Documents
Restek™
910150212
10 mm 3/Pk.
Pack of 3 for $194.00
Add to cart
06-611-652
View Documents
Restek™
910250210
10 mm 3/Pk.
Pack of 3 for $194.00
Add to cart
06-611-660
View Documents
Restek™
917950210
10 mm 3/Pk.
Pack of 3 for $194.00
Add to cart
06-611-662
View Documents
Restek™
918150212
10 mm 3/Pk.
Pack of 3 for $194.00
Add to cart
06-611-661
View Documents
Restek™
918150210
10 mm 3/Pk.
Pack of 3 for $194.00
Add to cart
06-611-650
View Documents
Restek™
910050212
10 mm 3/Pk.
Pack of 3 for $194.00
Add to cart
06-611-658
View Documents
Restek™
916550212
10 mm 3/Pk.
Pack of 3 for $196.00
Add to cart
06-719-067
View Documents
Restek™
910050222
2.1 mm 20 mm 2 Pack Silica This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
06-719-078
View Documents
Restek™
910350220
4 mm 20 mm Each C8 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
06-719-163
View Documents
Restek™
917450220
4 mm 20 mm Each C18 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
06-719-164
View Documents
Restek™
917450222
2.1 mm 20 mm 2 Pack C18 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
06-719-079
View Documents
Restek™
910350222
4.6 mm 20 mm 2 Pack C8 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
06-719-086
View Documents
Restek™
910650220
4 mm 20 mm Each Cyano This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
06-719-168
View Documents
Restek™
917550222
4.6 mm 20 mm 2 Pack IBD This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
06-719-071
View Documents
Restek™
910150222
2.1 mm 20 mm 2 Pack C1 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
06-719-087
View Documents
Restek™
910650222
2.1 mm 20 mm 2 Pack Cyano This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
06-719-171
View Documents
Restek™
917850220
4 mm 20 mm Each Aqueous C18 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
06-719-172
View Documents
Restek™
917850222
2.1 mm 20 mm 2 Pack Aqueous C18 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
06-719-075
View Documents
Restek™
910250222
2 mm 20 mm 2 Pack C4 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Particle Size: 5µm, spherical
  • Pore size: 100 angstrom
Specifications
Specifications
100Å
Spherical
2.5 to 8