Learn More
Restek™ Ultra HPLC Columns with Amino Column Packing, 3um Particle Size
Excellent choice for the analysis of simple sugars
Manufacturer:  Restek™ 9107363700
 View more versions of this product
Catalog No. 06-715-220
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
2%
No
150mm
3μm
80°C
3.2mm
With Trident Inlet Fitting
Aminopropylsilane
2.5 to 8
HPLC column