Restek™ Ultra HPLC Columns with Aqueous C18 Column Packing
Rugged, reversed-phase column with a well-balanced retention profile. Restek Ultra Aqueous C18 Columns can effectively retain more types of solutes than a conventional C18 and are ideal for multicomponent LC-MS analyses.
Manufacturer: Restek™ 9178332700
 View more versions of this product
Catalog No. 06-716-535
  Check Availability
Add to Cart
Related Products
Product Content Feedback