Learn More
Restek™ Ultra HPLC Columns with C1 Column Packing, 3um Particle Size
For selective and reliable HPLC applications
Manufacturer:  Restek™ 9101312700
 View more versions of this product
Catalog No. 06-714-793
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
5%
3μm
Trident™ inlet fitting
316 Stainless Steel
Spherical
2.5 to 8
80°C
L13
Ultra C1
2.1mm
0.31mL/min.
100mm
Silica
3μm
100Å
HPLC Column
HPLC