Learn More
Restek™ Ultra HPLC Columns with C1 Column Packing, 3um Particle Size
For selective and reliable HPLC applications
Manufacturer:  Restek™ 9101363700
 View more versions of this product
Catalog No. 06-714-823
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
5%
3.2mm
150mm
3μm
100Å
HPLC column
Ultra C1
With Trident Inlet Fitting
Spherical
2.5 to 8
80°C
L13