Restek™ Ultra HPLC Columns with C18 Column Packing, 3um Particle Size
Similar in pore size to Pinnacle II™ columns, but with greater retention
Manufacturer: Restek™ 9174333700
 View more versions of this product
Catalog No. 06-716-290
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Particle Shape Spherical
    Stationary Phase Ultra C18
    Film Thickness 3μm
    Includes Trident Inlet Fitting
  • Description & Specifications
Related Products
Product Content Feedback