Learn More
Restek™ Ultra HPLC Columns with C18 Column Packing, 3um Particle Size
Similar in pore size to Pinnacle II™ columns, but with greater retention
Manufacturer:  Restek™ 9174335
 View more versions of this product
Catalog No. 06-716-291
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
20%
4.6mm
3μm
Anilines, Barbiturates, Carbonyls, fat soluble vitamins, Fatty Acids, Glycerides, Phthalates, PTH Amino Acids, Steroids, other Acids
30mm
Silica
3μm
100Å
HPLC Column
HPLC
Ultra C18
Yes
1.5mL/min.
Without Trident Inlet Fitting
316 Stainless Steel
Spherical
2.5 to 8
80°C
L1
Ultra C18