Learn More
Restek™ Ultra HPLC Columns with C18 Column Packing, 5um Particle Size
Similar in pore size to Pinnacle II™ columns, but with greater retention
Manufacturer:  Restek™ 9174531
 View more versions of this product
Catalog No. 06-716-328
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
20%
1mm
5μm
Anilines, Barbiturates, Carbonyls, fat soluble vitamins, Fatty Acids, Glycerides, Phthalates, PTH Amino Acids, Steroids, other Acids
Monomeric C18
4000psi
Spherical
2.5 to 8
10g
HPLC Column
HPLC
Ultra C18
Yes
0.05mL/min.
30mm
Stainless Steel
Type B Silica
5μm
100Å
80°C
L1
Ultra C18