Restek™ Ultra HPLC Columns with C18 Column Packing, Trident Inlet Fitting
Similar in pore size to Pinnacle II™ columns, but with greater retention
Manufacturer: Restek™ 9174573700
 View more versions of this product
Catalog No. 06-716-354
  Check Availability
Add to Cart
    Specifications
    Length (Metric) 250mm
    Diameter (Metric) Inner 3.2mm
    Includes Trident™ inlet fitting
  • Description & Specifications
Related Products
Product Content Feedback