Learn More
Restek™ Ultra HPLC Columns with C8 Column Packing, 3um Particle Size
Excellent for a wide range of analyses
Manufacturer:  Restek™ 9103355700
 View more versions of this product
Catalog No. 06-714-976
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
12%
4.6mm
3μm
Wide range of compounds from Acidic through slightly basic
50mm
Silica
3μm
100Å
HPLC Column
HPLC
Ultra C8
Yes
1.5mL/min.
Trident™ inlet fitting
316 Stainless Steel
Spherical
2.5 to 8
80°C
L7
Ultra C8