Learn More
Restek™ Ultra HPLC Columns with C8 Column Packing, 3um Particle Size
Excellent for a wide range of analyses
Manufacturer:  Restek™ 9103365700
 View more versions of this product
Catalog No. 06-714-983
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
12%
4.6mm
Trident Inlet Fitting
Monomeric C8
3μm
C8, octylsilane
80°C
L7
Ultra C8
Yes
150mm
Spherical
2.5 to 8
100Å
HPLC column