Learn More
Restek™ Ultra HPLC Columns with C8 Column Packing, 3um Particle Size
Excellent for a wide range of analyses
Manufacturer:  Restek™ 9103351
 View more versions of this product
Catalog No. 06-714-970
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
12%
1mm
3μm
Wide range of compounds from Acidic through slightly basic
316 Stainless Steel
Spherical
2.5 to 8
80°C
L7
Ultra C8
Ultra C8
Yes
0.07mL/min.
50mm
Silica
3μm
100Å
HPLC Column
HPLC