Learn More
Restek™ Ultra HPLC Columns with C8 Column Packing, 5um Particle Size
Excellent for a wide range of analyses
Manufacturer:  Restek™ 9103563700
 View more versions of this product
Catalog No. 06-715-019
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
12%
3.2mm
5μm
Wide range of compounds from Acidic through slightly basic
150mm
Stainless Steel
Type B Silica
5μm
100Å
80°C
L7
Ultra C8
Ultra C8
Yes
0.5mL/min.
Trident™ inlet fitting
Monomeric C8
4000psi
Spherical
2.5 to 8
10g
HPLC Column
HPLC