Learn More
Restek™ Ultra HPLC Columns with Cyano Column Packing, 3um Particle Size
Similar in pore size to Pinnacle II™ columns, but with greater retention
Manufacturer:  Restek™ 9106365700
 View more versions of this product
Catalog No. 06-715-143
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
8%
4.6mm
Trident Inlet Fitting
Cyanopropylsilane
3μm
100Å
HPLC column
Ultra Cyano
Yes
150m
Spherical
2.5 to 8
80°C
L10