Restek™ Ultra HPLC Columns with IBD Column Packing, 3um Particle Size
Similar in pore size to Pinnacle II™ columns, but with greater retention
Manufacturer: Restek™ 9175352700
 View more versions of this product
Catalog No. 06-716-380
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Pore Size 100Å
    pH 2.5 to 8
    Type HPLC Column
    Length (Metric) 50mm
    Diameter (Metric) Inner 2.1mm
  • Description & Specifications
Related Products
Product Content Feedback