Restek™ Ultra HPLC Columns with IBD Column Packing, 3um Particle Size
Similar in pore size to Pinnacle II™ columns, but with greater retention
Manufacturer: Restek™ 9175335700
 View more versions of this product
Catalog No. 06-716-377
  Check Availability
Add to Cart
    Specifications
    Pore Size 100Å
    pH 2.5 to 8
    Type HPLC Column
    Length (Metric) 30mm
    Diameter (Metric) Inner 4.6mm
  • Description & Specifications
Related Products
Product Content Feedback