Learn More
Restek™ Ultra HPLC Columns with IBD Column Packing, 3um Particle Size
Similar in pore size to Pinnacle II™ columns, but with greater retention
Manufacturer:  Restek™ 9175312700
 View more versions of this product
Catalog No. 06-716-359
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
2.1mm
2.5 to 8
HPLC Column
100mm
100Å