Learn More
Restek™ Ultra HPLC Columns with Silica Column Packing, 3um Particle Size
Similar in pore size to Pinnacle II™ columns, but with greater retention
Manufacturer:  Restek™ 9100351
 View more versions of this product
Catalog No. 06-714-733
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Ultra Silica
No
0.07mL/min.
50mm
Trace Amounts
Spherical
2.5 to 8
80°C
L3
Ultra Silica
1mm
3μm
Normal phase applications with high density bondings
316 Stainless Steel
Silica
3μm
100Å
HPLC Column
HPLC