Learn More
Restek™ Ultra II Biphenyl HPLC Guard Cartridges
For use with Ultra II Biphenyl HPLC columns
Manufacturer:  Restek™ 960950212
 View more versions of this product
Catalog No. 06-738-460
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Hypersil™ HPLC column
L11
Spherical
100Å
10mm
80°C
15%
Yes
3 or 5μm
2.5 to 8
2.1mm