Restek™ Uniliner™ Inlet Liners: Open-Top
For Agilent GCs equipped with split/splitless inlets.
Manufacturer: Restek™ 20316211.5
 View more versions of this product
Catalog No. 06-717-689
  Check Availability
Add to Cart
    Specifications
    Type Open-top Liner
    Quantity Ea.
    Material Borosilicate Glass
    Diameter (Metric) Inner 5mm
  • Description & Specifications
Related Products
Product Content Feedback