Restek™ Uniliner Liners for 1/4-Inch Packed Injection Port Conversion
Manufacturer: Restek™ 20903
 View more versions of this product
Catalog No. 06-718-149
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Part Not Found  Please contact customer service at 1-800-766-7000 with error code:999. 
    Specifications
    Diameter (Metric) Inner 0.53mm
    For Use With (Equipment) 0.53mm I.D. On-Column injection mode, 0.25 in. Packed injection port conversion, small buffer volume chamber 60mm L, for ≤2μL injections
    Quantity 5/Pack
    Product Type Uniliner™ liner
  • Description & Specifications
Related Products
Product Content Feedback