Restek™ Uniliner Liners for 1/4-Inch Packed Injection Port Conversion
For 0.25 in. packed injection port conversion.
Manufacturer: Restek™ 20234
 View more versions of this product
Catalog No. 06-717-642
  Check Availability
Add to Cart
    Specifications
    Product Type Ferrules
    For Use With (Equipment) Uniliner™ sleeve adapters
    Quantity 5/Pack
    • Reduces solvent tailing
    • 0.53mm ID version can be used in the direct (DI) or on-column (OC) injection mode
    • Incorporates a gentle taper that seals the column and reduces dead volume in direct injection mode
  • Description & Specifications
Related Products
Product Content Feedback