Restek™ Uniliner Liners for 1/4-Inch Packed Injection Port Conversion
Manufacturer: Restek™ 20309
 View more versions of this product
Catalog No. 06-717-683
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Diameter (Metric) Inner 0.32/0.53mm
    Diameter (Metric) Outer 0.5cm
    For Use With (Equipment) 0.32 and 0.53mm I.D. Column direct injection only, 0.25 in. Packed injection port conversion, Large buffer volume chamber 85mm L, for ≤4μL injections
    Quantity 5/Pack
    Product Type Uniliner™ liner
  • Description & Specifications
Related Products
Product Content Feedback