Restek™ Uniliner Liners for 1/4-Inch Packed Injection Port Conversion
Manufacturer: Restek™ 20308
 View all options for this product
Catalog No. 06-710-562
  Check Availability
Add to Cart
    Diameter (Metric) Inner 0.32/0.53mm
    Diameter (Metric) Outer 0.5cm
    For Use With (Equipment) 0.32 and 0.53mm I.D. Column direct injection only, 0.25 in. Injection ports to troubleshoot purge and trap units, Large buffer volume chamber 85mm long, for ≤4μL injections
    Quantity Each
    Product Type Uniliner™ liner
  • Description & Specifications
Related Products
Product Content Feedback