Restek™ Veriflo Flow Controllers
Available in Siltek-treated and stainless steel versions.
Manufacturer: Restek™ 24259
 View more versions of this product
Catalog No. 06-712-259
  Check Availability
Add to Cart
    Specifications
    Material Siltek™ Treated
    Size (English) Orifice 0.006 in.
    Size (Metric) Orifice 0.015cm
    Product Type Veriflo™ SC423XL Flow Controller
    • Available with or without a critical orifice
    • Interchangeable critical orifice
    Order Info
    Order orifices for alternate sampling times, or replacement orifices, separately.
  • Description & Specifications
Related Products
Product Content Feedback