Restek™ Viva C4 HPLC Prep Columns, 5um Particle Size
Highly base-deactivated wide pore packing that exhibits excellent peak shape for a wide range of compounds. Less retention in reversed phase assays than Viva C18 or Viva C8.
Manufacturer: Restek™ 9512578
 View more versions of this product
Catalog No. 06-722-444
  Check Availability
Add to Cart
Related Products
Product Content Feedback