Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Viva C4 HPLC Prep Columns, 5um Particle Size

Highly base-deactivated wide pore packing that exhibits excellent peak shape for a wide range of compounds. Less retention in reversed phase assays than Viva C18 or Viva C8.

Manufacturer:  Restek™ 9512558

 View more versions of this product

Catalog No. 06-722-435


  Check Availability
Add to cart

Specifications

Description

  • Endcapped
  • pH range: 2.5 to 8
  • Temperature limit: 80°C
  • Particle size: 5µm

Description & Specifications

Specifications

3.5%
Prep
Yes
Dispersive interaction
Silica
5μm
Non Polar
1 Ea.
80°C
L26
Viva™ C4
Butyl Silane
21.2mm
5μm
50mm
Spherical
2.5 to 8
300Å
100m2/g
HPLC preparative column
Reverse phase HPLC
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit