Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Viva C8 HPLC Prep Columns
The C8 is a conventional monomeric octylsilane column offering a shorter alkyl chain to provide less hydrophobic retention and improved basic peak shape over a traditional C18 phase.
Supplier: Restek™ 9513518
Description
- Like the C18, this general-purpose Restek C8 is suitable for a wide range of compounds from acidic through slightly basic
- Easy scale-up from Restek analytical columns
- Popular bonded phases
Specifications
Octylsilane | |
21.2 mm | |
Proteins and Other Higher Molecular Weight Compounds, Less Retentive Than C18 Phase | |
Monomeric C8 | |
5 μm | |
300 Å | |
100 m²/g | |
L7 | |
Silica | |
Reversed Phase | |
Reversed Phase | |
Preparative Column |
Preparative Column | |
5 μm | |
100 mm | |
Spherical | |
2.5 to 8 | |
1 Ea. | |
80°C | |
C8 | |
5% | |
Yes | |
Silica |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction