Learn More
Restek™ Viva C8 HPLC Prep Columns
The C8 is a conventional monomeric octylsilane column offering a shorter alkyl chain to provide less hydrophobic retention and improved basic peak shape over a traditional C18 phase.
Manufacturer:  Restek™ 9513569
 View more versions of this product
Catalog No. 06-722-358
  Check Availability
Add to cart
Specifications
Description
  • Like the C18, this general-purpose Restek C8 is suitable for a wide range of compounds from acidic through slightly basic
  • Easy scale-up from Restek analytical columns
  • Popular bonded phases
Description & Specifications
Specifications
5%
Prep
30mm
5μm
150mm
Silica
Spherical
2.5 to 8
300Å
100m2/g
HPLC preparative column
Viva™ C8
Octylsilane
Viva™ C8
Yes
Proteins and other higher molecular weight compounds, less retentive than C18 phase
Monomeric C8
Silica
5μm
C8
1 Ea.
80°C
L7