Restek™ Viva C8 HPLC Prep Columns

Manufacturer: Restek™ 9513577

 View more versions of this product

Catalog No. 06-722-359

  Check Availability
Add to Cart

    Specifications

    Particle Shape Spherical
    Stationary Phase Viva C8
    Type HPLC preparative column
    Length (Metric) 250mm
    Material Silica
    Support Material Silica
    Diameter (Metric) Inner 10mm
    Phase C8
    Column Type Viva C8
    Packing Material Viva C8
    Particle Size 5μm
    Pore Size 300Å
    pH 2.5 to 8
    For Use With Proteins and other higher molecular weight compounds, less retentive than C18 phase
    Surface Area 100m2/g
    Film Thickness 5μm
    Carbon Load 5%
    Ligand Type Monomeric C8
    Column Format Prep
    Column Chemistry Octylsilane
    Max. Temperature 80°C
    USP Type L7
    Endcapped Yes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit