Restek™ Viva C8 HPLC Prep Columns

Manufacturer: Restek™ 9513568

 View more versions of this product

Catalog No. 06-722-357

  Check Availability
Add to Cart

    Specifications

    Max. Temperature 80°C
    Column Type Viva C8
    Packing Material Viva C8
    Particle Size 5μm
    Stationary Phase Viva C8
    Surface Area 100m2/g
    Type HPLC preparative column
    Length (Metric) 150mm
    Material Silica
    Support Material Silica
    Diameter (Metric) Inner 21.2mm
    Ligand Type Monomeric C8
    Column Chemistry Octylsilane
    USP Type L7
    Endcapped Yes
    Particle Shape Spherical
    Pore Size 300Å
    pH 2.5 to 8
    For Use With Proteins and other higher molecular weight compounds, less retentive than C18 phase
    Film Thickness 5μm
    Carbon Load 5%
    Phase C8
    Column Format Prep

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit