Restek™ Viva™ HPLC Columns with C18 Column Packing, 3µm Particle Size

Ideal for separating peptides or proteins.

Manufacturer: Restek™ 9514331

 View more versions of this product

Catalog No. 06-739-193

  Check Availability
Add to Cart

    Specifications

    Flow Rate 0.07mL/min.
    Technique HPLC
    Column Type Viva™ C18
    Packing Material Viva™ C18
    Particle Shape Spherical
    Pore Size 300Å
    pH 2.5 to 7.5
    For Use With Proteins and other higher molecular weight compounds
    Stationary Phase Viva™ C18
    Film Thickness 3μm
    Type HPLC Column
    Length (Metric) 30mm
    Material 316 Stainless Steel
    Diameter (Metric) Inner 1mm
    Carbon Load 9%
    Temperature (Metric) 80°C
    USP Type L1
    Endcapped Yes
    Particle Size 3μm

    • Rugged, spherical particles with 300 angstrom pore size
    • High proportion of pore/surface area available to large molecules
    • General-purpose Restek C18 is a conventional monomeric octadecylsilane column
    • Particle Size: 3µm
    • USP Phase Code: L1

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit