Restek™ Viva™ HPLC Columns with C18 Column Packing, 5µm Particle Size

Wide bore silica columns excellent for general-purpose applications and separating peptides or proteins

Manufacturer: Restek™ 9514521

 View more versions of this product

Catalog No. 06-739-208

  Check Availability
Add to Cart

    Specifications

    Max. Pressure 4000psi
    Particle Size 5μm
    pH 2.5 to 10
    For Use With Analyzing large molecules and biomolecules
    Type HPLC Column
    Support Material Silica
    Flow Rate 0.05mL/min.
    Particle Shape Spherical
    Pore Size 300Å
    Material Stainless Steel
    Diameter (Metric) Inner 1mm
    Ligand Type Monomeric C18
    Temperature (Metric) 80°C
    Packing Material Viva™ C18
    Stationary Phase Viva™ C18
    Film Thickness 5μm
    Length (Metric) 200mm
    Carbon Load 9%
    Max. Temperature 80°C
    USP Type L1
    Endcapped Yes
    Column Type Viva™ C18
    Technique HPLC

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit