Restek™ Viva HPLC Prep Columns with C18 Column Packing, 5um Particle Size
Highly base-deactivated wide pore packing that exhibits excellent peak shape for a wide range of compounds. Excellent general-purpose column for analyzing large molecules and biomolecules.
Manufacturer: Restek™ 9514569
 View more versions of this product
Catalog No. 06-722-370
  Check Availability
Add to Cart
Related Products
Product Content Feedback