Learn More
Restek™ Viva HPLC Prep Columns with C18 Column Packing, 5um Particle Size
Highly base-deactivated wide pore packing that exhibits excellent peak shape for a wide range of compounds. Excellent general-purpose column for analyzing large molecules and biomolecules.
Manufacturer:  Restek™ 9514559
 View more versions of this product
Catalog No. 06-722-364
  Check Availability
Add to cart
Specifications
Description
  • Endcapped
  • Easy scale-up
  • Spherical silica
  • Pack with popular bonded phases like Ultra, Allure, Viva
Description & Specifications
Specifications
1 Ea.
Spherical
Viva C18 HPLC prep column
100m2/g
L1
9%
300Å
5μm
Monomeric
C18
2.5 to 8
C18
80°C
Viva C18 HPLC Prep Column
50mm
Yes
30mm