Learn More
Restek™ WISP 48 Screw-Thread Step Vial
Vials only
Manufacturer:  Restek™ 24655
 View more versions of this product
Catalog No. 06-718-988
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
WISP™ 48 Step™ Vial
4mL
Screw cap
1000 Pack
Yes
Yes
13-425
Clear
45mm
15mm
1.77 in.
4.0mL