Learn More
Promega Restriction Enzymes - Cla I Blue/White Cloning and Genome Qualified (C)
Capable of digesting DNA in 15 minutes or less.
Supplier: Promega R6551
Description
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq(R) Green Master Mix
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
- Genome qualified to ensure optimal performance in genomic analysis applications
Specifications
10 U/μL | |
37°C | |
Capable of digesting DNA in <15 min. | |
-30°C to -10°C | |
AT.CGAT | |
Liquid |
1 x 1mL 10X Buffer C, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 500U ClaI, 1 x 0.25mL Multi-Core™ 10X Buffer | |
7.9 | |
10mM Tris HCl (pH 7.4), 300mM Sodium Glutamate, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol | |
500 U | |
ClaI |