Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Restriction Enzymes - Hinc II Blue/White Cloning Qualified (B)
Active and capable of digestion directly in GoTaq™ Green Master Mix. Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications. (GT(T/C)/(A/G)AC CA(A/G)\(T/C)TG)
Specifications
Format | Liquid |
---|---|
Concentration | 10 U/μL |
Incubator Temperature | 37°C |
For Use With (Application) | Provides a higher level of quality control for Enzymes used in cloning applications |
Storage Buffer | 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol |
Catalog Number | Mfr. No. | Quantity | Components | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Components | Price | Quantity & Availability | ||||
PRR6035
|
Promega
R6035 |
1000 U | 1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 1000U HincII, 1 x 0.25mL Multi-Core™ 10X Buffer | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- Active and capable of digestion directly in GoTaq(R) Green Master Mix
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.
Specifications
Liquid | |
37°C | |
10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol | |
GT(T/C).(A/G)AC CA(A/G).(T/C)TG |
10 U/μL | |
Provides a higher level of quality control for Enzymes used in cloning applications | |
-30°C to -10°C | |
HincII |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title