Learn More
Echelon Biosciences Research Labs RESVERATROL 100 mg
SDP
Supplier:  Echelon Biosciences Research Labs B0021100MG
Encompass
Resveratrol is a class IA PI3K inhibitor, activates SIRT1, and displays antiproliferative effect on human breast epithelial cells. By depolarization of mitochondrial membrane and caspase-9 activation resveratrol induces apoptosis. In addition, Resveratrol inhibits ribonucleotide reductase and DNA synthesis, cyclooxygenase-1, PKD, and eicosanoid synthesis. This item is non returnable
Catalog No. 50-720-001