Learn More
Echelon Biosciences Research LabsSupplier Diversity Partner RESVERATROL 50 mg  DFS Item
Manufacturer:  Echelon Biosciences Research LabsSupplier Diversity Partner B002150MG
Resveratrol is a class IA PI3K inhibitor, activates SIRT1, and displays antiproliferative effect on human breast epithelial cells. By depolarization of mitochondrial membrane and caspase-9 activation resveratrol induces apoptosis. In addition, Resveratrol inhibits ribonucleotide reductase and DNA synthesis, cyclooxygenase-1, PKD, and eicosanoid synthesis.
Catalog No. 50-720-000