Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ret Mouse anti-Human, Clone: RET/2599, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Ret
Clone RET/2599
Concentration 0.2 mg/ml
Dilution ELISA 2-4 ug/ml, Protein Array
Applications ELISA, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122302
View Documents
Novus Biologicals
NBP30741420UG
20 μg
Each for $138.37
Add to cart
 
NB122301
View Documents
Novus Biologicals
NBP30741410UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

Ret Monoclonal antibody specifically detects Ret in Human samples. It is validated for ELISA, Protein Array
Specifications

Specifications

Ret
0.2 mg/ml
ELISA, Peptide Array
Purified
RUO
Human
CDHF12MTC1, CDHR16, EC 2.7.10, Hirschsprung disease 1, Hirschsprung disease), MEN2A, MEN2B, multiple endocrine neoplasia and medullary thyroid carcinoma 1, ret proto-oncogene, RET transforming sequence
Recombinant fragment (around aa 702-848) of human Ret protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
RET/2599
ELISA 2-4 ug/ml, Protein Array
Unconjugated
Mouse
Cancer, Oncogenes, Phospho Specific, Protein Kinase, Protein Phosphatase, Signal Transduction, Tumor Suppressors
10 mM PBS with 0.05% BSA
5979
IgG2b Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit